Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PANK1 Full-length ORF (NP_683879.1, 1 a.a. - 373 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Specifications

Name PANK1 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 68.1kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-955-652
View Documents
Abnova™
H00053354P01L
25 ug N/A
89-955-651
View Documents
Abnova™
H00053354P01S
10 ug N/A
Description

Description

This gene encodes a protein belonging to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA. Alternative splicing has been observed at this locus and three variants, each encoding a distinct isoform, have been identified. [provided by RefSeq]

Sequence: MKLINGKKQTFPWFGMDIGGTLVKLVYFEPKDITAEEEQEEVENLKSIRKYLTSNTAYGKTGIRDVHLELKNLTMCGRKGNLHFIRFPSCAMHRFIQMGSEKNFSSLHTTLCATGGGAFKFEEDFRMIADLQLHKLDELDCLIQGLLYVDSVGFNGKPECYYFENPTNPELCQKKPYCLDNPYPMLLVNMGSGVSILAVYSKDNYKRVTGTSLGGGTFLGLCCLLTGCETFEEALEMAAKGDSTNVDKLVKDIYGGDYERFGLQGSAVASSFGNMMSKEKRDSISKEDLARATLVTITNNIGSIARMCALNENIDRVVFVGNFLRINMVSMKLLAYAMDFWSKGQLKALFLEHEGYFGAVGALLELFKMTDDK
Specifications

Specifications

PANK1 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
68.1kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit