Learn More
Human Papilloma Virus, type 11, oncoprotein E7 Mouse anti-Human, DyLight 650, Clone: 711-13, 711-45, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11059903C
Catalog No. NB11059903C
Add to cart
Description
Description
HPV11 E7 Monoclonal specifically detects HPV11 E7 in Virus samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
Human Papilloma Virus, type 11, oncoprotein E7
711-13, 711-45
Western Blot, ELISA
Purified
Mouse
IgG1/IgG2a
0.2 ml
Store at 4C in the dark.
Monoclonal
Human
Western Blot, ELISA
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
Human papillomavirus E7 protein
human papilloma virus (HPV), type 11, oncoprotein E7 (711-13, 711-45)
Protein G purified
RUO
Primary
human papilloma virus (HPV), type 11, oncoprotein E7 (711-13, 711-45)