Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PAPSS1 Full-length ORF (AAH11392.1, 1 a.a. - 603 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009061P01L

 View more versions of this product

Catalog No. 89-955-688


Add to cart

Description

Description

Three-prime-phosphoadenosine 5-prime-phosphosulfate (PAPS) is the sulfate donor cosubstrate for all sulfotransferase (SULT) enzymes (Xu et al., 2000 [PubMed 10679223]). SULTs catalyze the sulfate conjugation of many endogenous and exogenous compounds, including drugs and other xenobiotics. In humans, PAPS is synthesized from adenosine 5-prime triphosphate (ATP) and inorganic sulfate by 2 isoforms, PAPSS1 and PAPSS2 (MIM 603005).[supplied by OMIM]

Sequence: MQRATNVTYQAHHVSRNKRGQVVGTRGGFRGCTVWLTGLSGAGKTTVSMALEEYLVCHGIPCYTLDGDNIRQGLNKNLGFSPEDREENVRRIAEVAKLFADAGLVCITSFISPYTQDRNNARQIHEGASLPFFEVFVDAPLHVCEQRDVKGLYKKARAGEIKGFTGIDSEYEKPEAPELVLKTDSCDVNDCVQQVVELLQERDIVPVDASYEVKELYVPENKLHLAKTDAETLPALKINKVDMQWVQVLAEGWATPLNGFMREREYLQCLHFDCLLDGGVINLSVPIVLTATHEDKERLDGCTAFALMYEGRRVAILRNPEFFEHRKEERCARQWGTTCKNHPYIKMVMEQGDWLIGGDLQVLDRVYWNDGLDQYRLTPTELKQKFKDMNADAVFAFQLRNPVHNGHALLMQDTHKQLLERGYRRPVLLLHPLGGWTKDDDVPLMWRMKQHAAVLEEGVLNPETTVVAIFPSPMMYAGPTEVQWHCRARMVAGANFYIVGRDPAGMPHPETGKDLYEPSHGAKVLTMAPGLITLEIVPFRVAAYNKKKKRMDYYDSEHHEDFEFISGTRMRKLAREGQKPPEGFMAPKAWTVLTEYYKSLEKA
Specifications

Specifications

AAH11392.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
94.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATPSK1|PAPSS|SK1
PAPSS1
Yes
wheat germ expression system
Liquid
9061
PAPSS1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQRATNVTYQAHHVSRNKRGQVVGTRGGFRGCTVWLTGLSGAGKTTVSMALEEYLVCHGIPCYTLDGDNIRQGLNKNLGFSPEDREENVRRIAEVAKLFADAGLVCITSFISPYTQDRNNARQIHEGASLPFFEVFVDAPLHVCEQRDVKGLYKKARAGEIKGFTGIDSEYEKPEAPELVLKTDSCDVNDCVQQVVELLQERDIVPVDASYEVKELYVPENKLHLAKTDAETLPALKINKVDMQWVQVLAEGWATPLNGFMREREYLQCLHFDCLLDGGVINLSVPIVLTATHEDKERLDGCTAFALMYEGRRVAILRNPEFFEHRKEERCARQWGTTCKNHPYIKMVMEQGDWLIGGDLQVLDRVYWNDGLDQYRLTPTELKQKFKDMNADAVFAFQLRNPVHNGHALLMQDTHKQLLERGYRRPVLLLHPLGGWTKDDDVPLMWRMKQHAAVLEEGVLNPETTVVAIFPSPMMYAGPTEVQWHCRARMVAGANFYIVGRDPAGMPHPETGKDLYEPSHGAKVLTMAPGLITLEIVPFRVAAYNKKKKRMDYYDSEHHEDFEFISGTRMRKLAREGQKPPEGFMAPKAWTVLTEYYKSLEKA
RUO
PAPSS1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit