Learn More
Abnova™ Human PARD6A Full-length ORF (NP_001032358.1, 1 a.a. - 345 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_001032358.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 50855 |
Molecular Weight (g/mol) | 63.7kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-955-709
|
Abnova™
H00050855P01L |
25 ug |
Each for $769.00
|
|
|||||
89-955-708
|
Abnova™
H00050855P01S |
10 ug |
Each for $499.00
|
|
|||||
Description
This gene is a member of the PAR6 family and encodes a protein with a PSD95/Discs-large/ZO1 (PDZ) domain and a semi-Cdc42/Rac interactive binding (CRIB) domain. This cell membrane protein is involved in asymmetrical cell division and cell polarization processes as a member of a multi-protein complex. The protein also has a role in the epithelial-to-mesenchymal transition (EMT) that characterizes the invasive phenotype associated with metastatic carcinomas. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: MARPQRTPARSPDSIVEVKSKFDAEFRRFALPRASVSGFQEFSRLLRAVHQIPGLDVLLGYTDAHGDLLPLTNDDSLHRALASGPPPLRLLVQKREADSSGLAFASNSLQRRKKGLLLRPVAPLRTRPPLLISLPQDFRQVSSVIDVDLLPETHRRVRLHKHGSDRPLGFYIRDGMSVRVAPQGLERVPGIFISRLVRGGLAESTGLLAVSDEILEVNGIEVAGKTLDQVTDMMVANSHNLIVTVKPANQRNNVVRGASGRLTGPPSAGPGPAEPDSDDDSSDLVIENRQPPSSNGLSQGPPCWDLHPGCRHPGTRSSLPSLDDQEQASSGWGSRIRGDGSGFSLSpecifications
NP_001032358.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MARPQRTPARSPDSIVEVKSKFDAEFRRFALPRASVSGFQEFSRLLRAVHQIPGLDVLLGYTDAHGDLLPLTNDDSLHRALASGPPPLRLLVQKREADSSGLAFASNSLQRRKKGLLLRPVAPLRTRPPLLISLPQDFRQVSSVIDVDLLPETHRRVRLHKHGSDRPLGFYIRDGMSVRVAPQGLERVPGIFISRLVRGGLAESTGLLAVSDEILEVNGIEVAGKTLDQVTDMMVANSHNLIVTVKPANQRNNVVRGASGRLTGPPSAGPGPAEPDSDDDSSDLVIENRQPPSSNGLSQGPPCWDLHPGCRHPGTRSSLPSLDDQEQASSGWGSRIRGDGSGFSL | |
RUO | |
PARD6A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
50855 | |
PARD6A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PAR-6A/PAR6/PAR6C/PAR6alpha/TAX40/TIP-40 | |
PARD6A | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.