Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Parechovirus 1/3 VP0 Antibody (HL2443) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325513
Description
Human Parechovirus 1/3 VP0 Monoclonal antibody specifically detects Human Parechovirus 1/3 VP0 in Virus samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
Human Parechovirus 1/3 VP0 | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Western Blot, Immunocytochemistry | |
HL2443 | |
Western Blot 1:500-1:10000, Immunocytochemistry/ Immunofluorescence | |
Capsid protein VP0, Human Parechovirus Capsid protein VP0, P1AB, Parechovirus A VP0, PeV-A VP0, Virion protein 0 | |
Full length of Human Parechovirus 3 VP0. | |
100 μL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction