Learn More
Raybiotech IncSupplier Diversity Partner Human Park7/DJ-1 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHPARK71
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 32 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Park7.
Catalog No. 50-194-9621