Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PASD1 Full-length ORF (AAH40301.1, 1 a.a. - 773 a.a.) Recombinant Protein with GST-tag at N-terminal (P02)

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00139135P02

Catalog No. 89-955-758


Add to cart

Description

Description

Sequence: MKMRGEKRRDKVNPKSSQRKLNWIPSFPTYDYFNQVTLQLLDGFMITLSTDGVIICVAENISSLLGHLPAEIVGKKLLSLLPDEEKDEVYQKIILKFPLLNSETHIEFCCHLKRGNVEHGDSSAYENVKFIVNVRDICNEFPVVFSGLFSSHLCADFAACVPQEDRLYLVGNVCILRTQLLQQLYTSKAVSDEAVLTQDSDEEPFVGELSSSQGQRGHTSMKAVYVEPAAAAAAAAISDDQIDIAEVEQYGPQENVHMFVDSDSTYCSSTVFLDTMPESPALSLQDFRGEPEVNPLYRADPVDLEFSVDQVDSVDQEGPMDQQDPENPVAPLDQAGLMDPVDPEDSVDLGAAGASAQPLQPSSPVAYDIISQELELMKKLKEQLEERTWLLHDAIQNQQNALELMMDHLQKQPNTLRHVVIPDLQSSEAVPKKQQKQHAGQVKRPLPHPKDVKCFCGLSLSNSLKNTGELQEPCVAFNQQQLVQQEQHLKEQQRQLREQLQQLREQRKVQKQKKMQEKKKLQEQKMQEKKKLQEQRRQKKKKLQERKKWQGQMLQKEPEEEQQKQQLQEQPLKHNVIVGNERVQICLQNPRDVSVPLCNHPVRFLQAQPIVPVQRAAEQQPSGFYQDENCGQQEDESQSFYPEAYQGPPVNQLPLIDTSNSEAISSSSIPQFPITSDSTISTLETPQDYIRLWQELSDSLGPVVQVNTWSCDEQGTLHGQPTYHQVQVSEVGVEGPPDPQAFQGPAAYQPDQMRSAEQTRLMPAEQRDSNKPC
Specifications

Specifications

AAH40301.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
113.8kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PASD1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
139135
PASD1 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKMRGEKRRDKVNPKSSQRKLNWIPSFPTYDYFNQVTLQLLDGFMITLSTDGVIICVAENISSLLGHLPAEIVGKKLLSLLPDEEKDEVYQKIILKFPLLNSETHIEFCCHLKRGNVEHGDSSAYENVKFIVNVRDICNEFPVVFSGLFSSHLCADFAACVPQEDRLYLVGNVCILRTQLLQQLYTSKAVSDEAVLTQDSDEEPFVGELSSSQGQRGHTSMKAVYVEPAAAAAAAAISDDQIDIAEVEQYGPQENVHMFVDSDSTYCSSTVFLDTMPESPALSLQDFRGEPEVNPLYRADPVDLEFSVDQVDSVDQEGPMDQQDPENPVAPLDQAGLMDPVDPEDSVDLGAAGASAQPLQPSSPVAYDIISQELELMKKLKEQLEERTWLLHDAIQNQQNALELMMDHLQKQPNTLRHVVIPDLQSSEAVPKKQQKQHAGQVKRPLPHPKDVKCFCGLSLSNSLKNTGELQEPCVAFNQQQLVQQEQHLKEQQRQLREQLQQLREQRKVQKQKKMQEKKKLQEQKMQEKKKLQEQRRQKKKKLQERKKWQGQMLQKEPEEEQQKQQLQEQPLKHNVIVGNERVQICLQNPRDVSVPLCNHPVRFLQAQPIVPVQRAAEQQPSGFYQDENCGQQEDESQSFYPEAYQGPPVNQLPLIDTSNSEAISSSSIPQFPITSDSTISTLETPQDYIRLWQELSDSLGPVVQVNTWSCDEQGTLHGQPTYHQVQVSEVGVEGPPDPQAFQGPAAYQPDQMRSAEQTRLMPAEQRDSNKPC
RUO
PASD1
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit