Learn More
Invitrogen™ Human Pax-4 ELISA Kit
ELISA
$666.00 - $3285.00
Specifications
Accession Number | KPD; MGC129960; MODY9; paired box 4; paired box gene 4; Paired box protein Pax-4; paired domain gene 4; Pax4; Pax-4 |
---|---|
Assay Range | 0.14 to 30 ng/mL |
Assay Sensitivity | 0.14 ng/mL |
Conjugate | HRP |
Product Type | ELISA |
Description
Human Pax-4 quantitates human Pax-4 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Pax-4.
This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The paired box 4 gene is involved in pancreatic islet development and mouse studies have demonstrated a role for this gene in differentiation of insulin-producing beta cells.Specifications
KPD; MGC129960; MODY9; paired box 4; paired box gene 4; Paired box protein Pax-4; paired domain gene 4; Pax4; Pax-4 | |
0.14 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
PAX4 | |
<10 | |
HRP | |
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
PAX4 | |
4 hrs 45 mins |
0.14 to 30 ng/mL | |
HRP | |
Plasma, Serum, Supernatant | |
5078 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.