Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PC-LKC Partial ORF (NP_060145, 210 a.a. - 318 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Specifications

Name PC-LKC (Human) Recombinant Protein (Q01)
Recombinant Yes
For Use With (Application) Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Molecular Weight 37.73kDa
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-956-043
View Documents
Abnova™
H00054825Q01L
25 ug N/A
89-956-042
View Documents
Abnova™
H00054825Q01S
10 ug N/A
Description

Description

This gene is a member of the protocadherin family, which represents a subset of the larger cadherin superfamily. The members of the protocadherin family encode non-classical cadherins that function as calcium-dependent cell-cell adhesion molecules. The kidney-expressed gene product has a signal peptide, nine cadherin repeat domains and a unique cytoplasmic region. This protocadherin represents a new candidate for tumor suppression. [provided by RefSeq]

Sequence: GGMYHNTFTIQCSLPVFLSISVVDQPDLDPQFVREFYSASVAEDAAKGTSVLTVEAVDGDKGINDPVIYSISYSTRPGWFDIGADGVIRVNGSLDREQLLEADEEVQLQ
Specifications

Specifications

PC-LKC (Human) Recombinant Protein (Q01)
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
37.73kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit