Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PCDH11X Partial ORF (NP_061850, 1228 a.a. - 1336 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00027328Q01L

 View more versions of this product

Catalog No. 89-955-857


Add to cart

Description

Description

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The encoded protein consists of an extracellular domain containing 7 cadherin repeats, a transmembrane domain and a cytoplasmic tail that differs from those of the classical cadherins. The gene is located in a major X/Y block of homology and its Y homolog, despite divergence leading to coding region changes, is the most closely related cadherin family member. The protein is thought to play a fundamental role in cell-cell recognition essential for the segmental development and function of the central nervous system. Transcripts arising from alternative splicing encode isoforms with variable cytoplasmic domains. [provided by RefSeq]

Sequence: AQASALCYSPPLAQAAAISHSSPLPQVIALHRSQAQSSVSLQQGWVQGADGLCSVDQGVQGSATSQFYTMSERLHPSDDSIKVIPLTTFTPRQQARPSRGDSPIMEEHP
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AQASALCYSPPLAQAAAISHSSPLPQVIALHRSQAQSSVSLQQGWVQGADGLCSVDQGVQGSATSQFYTMSERLHPSDDSIKVIPLTTFTPRQQARPSRGDSPIMEEHP
RUO
PCDH11X
Wheat Germ (in vitro)
GST
NP_061850
Liquid
27328
PCDH11X (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PCDH-X/PCDH11/PCDHX/PCDHY
PCDH11X
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit