Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human PCDH19 Partial ORF (NP_065817.1, 241 a.a. - 340 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00057526Q01S
Description
Protocadherins form a subfamily of calcium-dependent cell-cell adhesion molecules in the cadherin superfamily. Protocadherin-10 belongs to a subclass of protocadherins that share a highly conserved 17-amino acid cytoplasmic motif (Wolverton and Lalande, 2001 [PubMed 11549318]). PCDH19 belongs to the delta-2 protocadherin subclass of the cadherin superfamily (Dibbens et al., 2008) [PubMed 18469813].[supplied by OMIM]
Sequence: RLVPRDVEETDKMNVVSCSSLTSSLNYFDYHQQTLPLGCRRSESTFLNVENQNTRNTSANHIYHHSFNSQGPQQPDLIINGAPLPETENYSFDSNYVNSRSpecifications
NP_065817.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
RLVPRDVEETDKMNVVSCSSLTSSLNYFDYHQQTLPLGCRRSESTFLNVENQNTRNTSANHIYHHSFNSQGPQQPDLIINGAPLPETENYSFDSNYVNSR | |
RUO | |
PCDH19 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57526 | |
PCDH19 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686P1843/EFMR/KIAA1313 | |
PCDH19 | |
Recombinant | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction