Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PCDH19 Partial ORF (NP_065817.1, 241 a.a. - 340 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00057526Q01L

 View more versions of this product

Catalog No. 89-955-861


Add to cart

Description

Description

Protocadherins form a subfamily of calcium-dependent cell-cell adhesion molecules in the cadherin superfamily. Protocadherin-10 belongs to a subclass of protocadherins that share a highly conserved 17-amino acid cytoplasmic motif (Wolverton and Lalande, 2001 [PubMed 11549318]). PCDH19 belongs to the delta-2 protocadherin subclass of the cadherin superfamily (Dibbens et al., 2008) [PubMed 18469813].[supplied by OMIM]

Sequence: RLVPRDVEETDKMNVVSCSSLTSSLNYFDYHQQTLPLGCRRSESTFLNVENQNTRNTSANHIYHHSFNSQGPQQPDLIINGAPLPETENYSFDSNYVNSR
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue
RLVPRDVEETDKMNVVSCSSLTSSLNYFDYHQQTLPLGCRRSESTFLNVENQNTRNTSANHIYHHSFNSQGPQQPDLIINGAPLPETENYSFDSNYVNSR
RUO
PCDH19
Wheat Germ (in vitro)
GST
NP_065817.1
Liquid
57526
PCDH19 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686P1843/EFMR/KIAA1313
PCDH19
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit