Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PCDH19 Partial ORF (NP_065817.1, 241 a.a. - 340 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$398.50 - $1083.00

Specifications

Name PCDH19 (Human) Recombinant Protein (Q01)
Recombinant Yes
For Use With (Application) Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Molecular Weight 36.63kDa
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-955-861
View Documents
Abnova™
H00057526Q01L
25 ug
Each for $1,083.00
Add to cart
 
89-955-860
View Documents
Abnova™
H00057526Q01S
10 ug
Each for $398.50
Add to cart
 
Description

Description

Protocadherins form a subfamily of calcium-dependent cell-cell adhesion molecules in the cadherin superfamily. Protocadherin-10 belongs to a subclass of protocadherins that share a highly conserved 17-amino acid cytoplasmic motif (Wolverton and Lalande, 2001 [PubMed 11549318]). PCDH19 belongs to the delta-2 protocadherin subclass of the cadherin superfamily (Dibbens et al., 2008) [PubMed 18469813].[supplied by OMIM]

Sequence: RLVPRDVEETDKMNVVSCSSLTSSLNYFDYHQQTLPLGCRRSESTFLNVENQNTRNTSANHIYHHSFNSQGPQQPDLIINGAPLPETENYSFDSNYVNSR
Specifications

Specifications

PCDH19 (Human) Recombinant Protein (Q01)
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
36.63kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit