Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PCDHA2 Full-length ORF (AAH03126.1, 1 a.a. - 808 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$390.00 - $1083.00

Specifications

Name PCDHA2 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 113.5kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-955-887
View Documents
Abnova™
H00056146P01L
25 ug
Each for $1,083.00
Add to cart
 
89-955-886
View Documents
Abnova™
H00056146P01S
10 ug
Each for $390.00
Add to cart
 
Description

Description

This gene is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences. The tandem array of 15 N-terminal exons, or variable exons, are followed by downstream C-terminal exons, or constant exons, which are shared by all genes in the cluster. The large, uninterrupted N-terminal exons each encode six cadherin ectodomains while the C-terminal exons encode the cytoplasmic domain. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins that most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been observed and additional variants have been suggested but their full-length nature has yet to be determined. [provided by RefSeq]

Sequence: MASSIRRGRGAWTRLLSLLLLAAWEVGSGQLRYSVPEEAKHGTFVGRIAQDLGLELEELVPRLFRVASKRHGDLLEVNLQNGILFVNSRIDREELCGRSAECSIHVEVIVDRPLQVFHVEVEVKDINDNPPIFPMTVKTIRFPESRLLDSRFPLEGASDADIGVNALLSYKLSSSEFFFLDIQANDELSESLSLVLGKSLDREETAEVNLLLVATDGGKPELTGTVQILIKVLDVNDNEPTFAQSVYKVKLLENTANGTLVVKLNASDADEGPNSEIVYSLGSDVSSTIQTKFTIDPISGEIRTKGKLDYEEAKSYEIQVTATDKGTPSMSGHCKISLKLVDINDNTPEVSITSLSLPISENASLGTVIALITVSDRDSGTNGHVTCSLTPHVPFKLVSTFKNYYSLVLDSALDRESVSAYELVVTARDGGSPSLWATTSVSIEVADVNDNAPAFAQPEYTVFVKENNPPGCHIFTVSAWDADAQENALVSYSLVERRVGERALSSYVSVHAESGKVYALQPLDHEEVELLQFQVSARDAGVPPLGSNVTLQVFVLDENDNAPALLAPRAGTAAGAVSELVPWSVGAGHVVAKVRAVDADSGYNAWLSYELQLGTGSARIPFRVGLYTGEISTTRALDEADSPRHRLLVLVKDHGEPALTATATVLVSLVESGQAPKASSRAWVGAAGSEATLVDVNVYLIIAICAVSSLLVLTVLLYTALRCSVPPTEGARAPGKPTLVCSSAVGSWSYSQQRRQRVCSGEDPPKTDLMAFSPSLSQGPDSAEEKQLSESEYVGKIWNFNLQIQLAS
Specifications

Specifications

PCDHA2 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
113.5kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit