Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PCDHB15 Full-length ORF (AAH38797, 1 a.a. - 787 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name PCDHB15 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 112.31kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-955-928
View Documents
Abnova™
H00056121P01S
10 ug
Each for $460.50
Add to cart
 
89-955-929
View Documents
Abnova™
H00056121P01L
25 ug
Each for $709.50
Add to cart
 
Description

Description

This gene is a member of the protocadherin beta gene cluster, one of three related gene clusters tandemly linked on chromosome five. The gene clusters demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The beta cluster contains 16 genes and 3 pseudogenes, each encoding 6 extracellular cadherin domains and a cytoplasmic tail that deviates from others in the cadherin superfamily. The extracellular domains interact in a homophilic manner to specify differential cell-cell connections. Unlike the alpha and gamma clusters, the transcripts from these genes are made up of only one large exon, not sharing common 3' exons as expected. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins. Their specific functions are unknown but they most likely play a critical role in the establishment and function of specific cell-cell neural connections. [provided by RefSeq]

Sequence: MEPAGERFPEQRQVLILLLLLEVTLAGWEPRRYSVMEETERGSFVANLANDLGLGVGELAERGARVVSEDNEQGLQLDLQTGQLILNEKLDREKLCGPTEPCIMHFQVLLKKPLEVFRAELLVTDINDHSPEFPEREMTLKIPETSSLGTVFPLKKARDLDVGSNNVQNYNISPNSHFHVSTRTRGDGRKYPELVLDTELDREEQAELRLTLTAVDGGSPPRSGTVQILILVLDANDNAPEFVQALYEVQVPENSPVGSLVVKVSARDLDTGTNGEISYSLYYSSQEIDKPFELSSLSGEIRLIKKLDFETMSSYDLDIEASDGGGLSGKCSVSVKVLDVNDNFPELSISSLTSPIPENSPETEVALFRIRDRDSGENGKMICSIQDDVPFKLKPSVENFYRLVTEGALDRETRAEYNITITITDLGTPRLKTEQSITVLVSDVNDNAPAFTQTSYTLFVRENNSPALHIGSVRATDRDSGTNAQVTYSLLPPQDPHLPLTSLVSINTDNGHLFALQSLDYEALQAFEFRVGATDRGFPALSSEALVRVLVLDANDNSPFVLYPLQNGSAPCTELVPRAAEPGYLVTKVVAVDGDSGQNAWLSYQLLKATEPGLFGVWAHNGEVRTARLLSERDVAKHRLVVLVKDNGEPPRSATATLQVLLVDGFSQPYLPLPEAAPAQAQADSLTVYLVVALASVSSLFLFSVFLFVAVRLCRRSRAASVGRCSVPEGPFPGHLVDVSGTGTLSQSYQYEVCLTGGSESNDFKFLKPIFPNIVSQDSRRKSEFLE
Specifications

Specifications

PCDHB15 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
112.31kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit