Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PCDHB6 Full-length ORF (AAI52976.1, 1 a.a. - 794 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Supplier:  Abnova™ H00056130P01L

 View more versions of this product

Catalog No. 89-955-941


Add to Cart

Description

Description

This gene is a member of the protocadherin beta gene cluster, one of three related gene clusters tandemly linked on chromosome five. The gene clusters demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The beta cluster contains 16 genes and 3 pseudogenes, each encoding 6 extracellular cadherin domains and a cytoplasmic tail that deviates from others in the cadherin superfamily. The extracellular domains interact in a homophilic manner to specify differential cell-cell connections. Unlike the alpha and gamma clusters, the transcripts from these genes are made up of only one large exon, not sharing common 3' exons as expected. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins. Their specific functions are unknown but they most likely play a critical role in the establishment and function of specific cell-cell neural connections. [provided by RefSeq]

Sequence: MMQTKVQNKKRQVAFFILLMLWGEVGSESIQYSVLEETESGTFVANLTKDLGLRVGELASRGARVVFKGNRQHLQFDPQTHDLLLNEKLDREELCGSTEPCVLPFQVLLENPLQFFQASLRVRDINDHAPEFPAREMLLKISEITMPGKIFPLKMAHDLDTGSNGLQRYTISSNPHFHVLTRNRSEGRKFPELVLDKPLDREEQPQLRLTLIALDGGSPPRSGTSEIQIQVLDINDNVPEFAQELYEAQVPENNPLGSLVITVSARDLDAGSFGKVSYALFQVDDVNQPFEINAITGEIRLRKALDFEEIQSYDVDVEATDGGGLSGKCSLVVRVLDVNDNAPELTMSFFISLIPENLPEITVAVFSVSDADSGHNQQVICSIENNLPFLLRPSVENFYTLVTEGALDRESRAEYNITITVTDLGTPRLKTQQSITVQVSDVNDNVPAFTQTSYTLFVRENNSPALHIGSVSATDRDSGINAQVTYSLLPPQDPHLPLSSLVSINADNGHLFALRSLDYEALQSFEFRVGATDRGSPALSSEALVRLLVLDANDNSPFVLYPLQNGSAPCTELVPRAAEPGYLVTKVVAVDGDSGQNAWLSYQLLKATELGLFGVWAHNGEVRTARLLSERDAAKHRLVVLVKDNGEPPRSATATLHVLLVDGFSQPYLPLPEAAPAQAQADSLTVYLVVALASVSSLFLFSVLLFVAVRLCRRSRAASVGRYSVPEGPFPGHLVDVSGTGTLSQSYQYKVCLTGGSETNEFKFLKPIMPNFPPQGTEREMEETPTSRNSFPFS
Specifications

Specifications

AAI52976.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
114.29kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PCDH-BETA6
PCDHB6
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
56130
PCDHB6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MMQTKVQNKKRQVAFFILLMLWGEVGSESIQYSVLEETESGTFVANLTKDLGLRVGELASRGARVVFKGNRQHLQFDPQTHDLLLNEKLDREELCGSTEPCVLPFQVLLENPLQFFQASLRVRDINDHAPEFPAREMLLKISEITMPGKIFPLKMAHDLDTGSNGLQRYTISSNPHFHVLTRNRSEGRKFPELVLDKPLDREEQPQLRLTLIALDGGSPPRSGTSEIQIQVLDINDNVPEFAQELYEAQVPENNPLGSLVITVSARDLDAGSFGKVSYALFQVDDVNQPFEINAITGEIRLRKALDFEEIQSYDVDVEATDGGGLSGKCSLVVRVLDVNDNAPELTMSFFISLIPENLPEITVAVFSVSDADSGHNQQVICSIENNLPFLLRPSVENFYTLVTEGALDRESRAEYNITITVTDLGTPRLKTQQSITVQVSDVNDNVPAFTQTSYTLFVRENNSPALHIGSVSATDRDSGINAQVTYSLLPPQDPHLPLSSLVSINADNGHLFALRSLDYEALQSFEFRVGATDRGSPALSSEALVRLLVLDANDNSPFVLYPLQNGSAPCTELVPRAAEPGYLVTKVVAVDGDSGQNAWLSYQLLKATELGLFGVWAHNGEVRTARLLSERDAAKHRLVVLVKDNGEPPRSATATLHVLLVDGFSQPYLPLPEAAPAQAQADSLTVYLVVALASVSSLFLFSVLLFVAVRLCRRSRAASVGRYSVPEGPFPGHLVDVSGTGTLSQSYQYKVCLTGGSETNEFKFLKPIMPNFPPQGTEREMEETPTSRNSFPFS
RUO
PCDHB6
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit