Learn More
Abnova™ Human PCDHB6 Partial ORF (NP_061762, 118 a.a. - 217 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00056130Q01S
Description
This gene is a member of the protocadherin beta gene cluster, one of three related gene clusters tandemly linked on chromosome five. The gene clusters demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The beta cluster contains 16 genes and 3 pseudogenes, each encoding 6 extracellular cadherin domains and a cytoplasmic tail that deviates from others in the cadherin superfamily. The extracellular domains interact in a homophilic manner to specify differential cell-cell connections. Unlike the alpha and gamma clusters, the transcripts from these genes are made up of only one large exon, not sharing common 3' exons as expected. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins. Their specific functions are unknown but they most likely play a critical role in the establishment and function of specific cell-cell neural connections. [provided by RefSeq]
Sequence: ASLRVRDINDHAPEFPAREMLLKISEITMPGKIFPLKMAHDLDTGSNGLQRYTISSNPHFHVLTRNRSEGRKFPELVLDKPLDREEQPQLRLTLIALDGGSpecifications
NP_061762 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ASLRVRDINDHAPEFPAREMLLKISEITMPGKIFPLKMAHDLDTGSNGLQRYTISSNPHFHVLTRNRSEGRKFPELVLDKPLDREEQPQLRLTLIALDGG | |
RUO | |
PCDHB6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
56130 | |
PCDHB6 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PCDH-BETA6 | |
PCDHB6 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.