Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PCDHGA11 Full-length ORF (NP_114480.1, 1 a.a. - 837 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name PCDHGA11 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 118kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-955-954
View Documents
Abnova™
H00056105P01L
25 ug
Each for $709.50
Add to cart
 
89-955-953
View Documents
Abnova™
H00056105P01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes. [provided by RefSeq]

Sequence: MANRLQRGDRSRLLLLLCIFLGTLRGFRARQIRYSVPEETEKGSFVGNISKDLGLEPRELAKRGVRIVSRGKTQLFAVNPRSGSLITAGRIDREELCETVSSCFLNMELLVEDTLKIYGVEVEIIDINDNAPSFQEDEVEIKVSEHAIPGARFALPNARDPDVGVNSLQSYQLSPNNYFSLQLRGRTDGAKNPELVLEGSLDREKEAAHLLLLTALDGGDPIRKGAVPIRVVVLDVNDHIPMFTQSVYRVSVPENISSGTRVLMVNATDPDEGINGEVMYSFRNMESKASEIFQLDSQTGEVQVRGSLDFEKYRFYEMEIQGQDGGGLFTTTTMLITVVDVNDNAPEITITSSINSILENSPPGTVIALLNVQDQDSGENGQVSCFIPNHLPFKLEKTYGNYYKLITSRVLDRELVQSYNITLTATDQGSPPLSAETHVWLNVADDNDNPPVFPHSSYSAYIPENNPRGASIFSVTALDPDSKQNALVTYSLTDDTVQGVPLSSYVSINSNTGVLYALQSFDYEQFRDLELRVIARDSGDPPLSSNVSLSLFVLDQNDNAPEILYPALPTDGSTGVELAPRSAEPGYLVTKVVAVDKDSGQNAWLSYRLLKASEPGLFAVGEHTGEVRTARALLDRDALKQSLVVAVQDHGQPPLSATVTLTVAVADSIPEVLADLGSLESLANSETSDLSLYLVVAVAAVSCIFLVFVIVLLALRLWRWHKSRLLQASEGGLAGMPTSHFVGVDGVQAFLQTYSHEVSLIADSQKSHLIFPQPNYGDTLISQESCEKSEPLLIAEDSAIILGKCDPTSNQVRFISLPPNCWCLGTSLLRRCFLSLL
Specifications

Specifications

PCDHGA11 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
118kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit