Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PCDHGB2 Partial ORF (NP_061746, 94 a.a. - 199 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00056103Q01S

 View more versions of this product

Catalog No. 89-955-988


Add to cart

Description

Description

This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes. [provided by RefSeq]

Sequence: QICGKQPLCVLDFDTVAENPLNIFYIAVIVQDINDNTPLFKQTKINLKIGESTKPGTTFPLDPALDSDVGPNSLQRYHLNDNEYFDLAEKQTPDGRKYPELILKHS
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.4kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QICGKQPLCVLDFDTVAENPLNIFYIAVIVQDINDNTPLFKQTKINLKIGESTKPGTTFPLDPALDSDVGPNSLQRYHLNDNEYFDLAEKQTPDGRKYPELILKHS
RUO
PCDHGB2
Wheat Germ (in vitro)
GST
NP_061746
Liquid
56103
PCDHGB2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC126854/PCDH-GAMMA-B2
PCDHGB2
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit