Learn More
Abnova™ Human PCDHGC5 Full-length ORF (NP_115783.1, 1 a.a. - 878 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00056097P01S
Description
This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes. [provided by RefSeq]
Sequence: MGPKTLPQLAGKWQVLCMLSLCCWGWVSGQLRYSVVEESEPGTLVGNVAQDLGLKMTDLLSRRLQLGSEENGRYFSLSLMSGALAVNQKIDRESLCGASTSCLLPVQVVTEHPLELIRVEVEILDLNDNSPSFATPEREMRISESAASGARFPLDSAQDPDVGTNTVSFYTLSPNSHFSLNVKTLKDGKPFPELVLEQQLDREAQARHQLVLTAVDGGTPARSGTTLISVIVLDINDNAPTFQSSVLRVGIPENAPIGTLLLRLNATDPDEGTNGQLDYSFGDHTSEAVRNLFGLDPSSGAIHVLGPIDFEESRFYEIHARARDQGQPAMEGHCVIQVDVGDVNDNAPEVLLASLANPVLESTPVGTVVGLFNVRDRDSGRNGEVSLDISPDLPFQIKPSENHYSLLTSQPLDREATSHYIIELLASDAGSPSLHKHLTIRLNISDVNDNAPRFNQQLYTAYILENRPPGSLLCTVAASDPDTGDNARLTYSIVGNQVQGAPASSFVYVNPEDGRIFAQRTFDYELLQMLQIVVGVRDSGSPPLHANTSLHVFVLDENDNAPAVLHPRPDWEHSAPQRLPRSAPPGSLVTKVTAVDADAGHNAWLSYSLLPQSTAPGLFLVSTHTGEVRTARALLEDDSDTQQVVVLVRDNGDPSLSSTATVLLVLEDEDPEEMPKSSDFLIHPPERSDLTLYLIVALATVSLLSLVTFTFLSAKCLQGNADGDGGGGQCCRRQDSPSPDFYKQSSPNLQVSSDGTLKYMEVTLRPTDSQSHCYRTCFSPASDGSDFTFLRPLSVQQPTALALEPDAIRSRSNTLRERSQVRGSAPPRATPGGGTGEAARPHKGLNLHPLLSGRLGRWLRSTRFSGSLDRLRETRVADSpecifications
NP_115783.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
121.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10μg | |
RUO | |
PCDHGC5 | |
Wheat Germ (in vitro) | |
GST Tag | |
Liquid |
Protein Array, ELISA, Western Blot | |
56097 | |
Human PCDHGC5 Full-length ORF (NP_115783.1, 1 a.a. - 878 a.a.) Recombinant Protein with GST-tag at N-terminal | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at −80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC138286, MGC138288, PCDH-GAMMA-C5 | |
PCDHGC5 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.