Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PCLO Full-length ORF (AAH01304.2, 1 a.a. - 356 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00027445P01L

 View more versions of this product

Catalog No. 89-956-045


Add to cart

Description

Description

Synaptic vesicles dock and fuse in the active zone of the plasma membrane at chemical synapses. The presynaptic cytoskeletal matrix (PCM), which is associated with the active zone and is situated between synaptic vesicles, is thought to be involved in maintaining the neurotransmitter release site in register with the postsynaptic reception apparatus. The cycling of synaptic vesicles is a multistep process involving a number of proteins (see MIM 603215). Among the components of the PCM that orchestrate these events are Bassoon (BSN; MIM 604020), RIM (RBBP8; MIM 604124), Oboe, and Piccolo (PCLO).[supplied by OMIM]

Sequence: MKKFRVSLVSKVGKQKYVDLNMLSDSENSQHLELHEPPKAVDKAKSPGVDPKQLAAELQKVSLQQSPLVLSSVVEKGSHVHSGPTSAGSSSVPSPGQPGSPSVSKKKHGSSKPTDGTKVVSHPITGEIQLQINYDLGNLIIHILQARNLVPRDNNGYSDPFVKVYLLPGRGAEYKRRTKHVQKSLNPEWNQTVIYKSISMEQLKKKTLEVTVWDYDRFSSNDFLGEVLIDLSSTAHLDNTPRWYPLKEQTESIDHGKSHSSQSSQQSPKPSVIKSRSHGIFPDPSKDMQVPTIEKSHSSPGSSKSSSEGHLRSHGPSRSQSKTSVTQTHLEDAGAAIAAAEAAVQQLRIQPSKRRK
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
65.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACZ/DKFZp779G1236
PCLO
Yes
AAH01304.2
Liquid
27445
PCLO (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKKFRVSLVSKVGKQKYVDLNMLSDSENSQHLELHEPPKAVDKAKSPGVDPKQLAAELQKVSLQQSPLVLSSVVEKGSHVHSGPTSAGSSSVPSPGQPGSPSVSKKKHGSSKPTDGTKVVSHPITGEIQLQINYDLGNLIIHILQARNLVPRDNNGYSDPFVKVYLLPGRGAEYKRRTKHVQKSLNPEWNQTVIYKSISMEQLKKKTLEVTVWDYDRFSSNDFLGEVLIDLSSTAHLDNTPRWYPLKEQTESIDHGKSHSSQSSQQSPKPSVIKSRSHGIFPDPSKDMQVPTIEKSHSSPGSSKSSSEGHLRSHGPSRSQSKTSVTQTHLEDAGAAIAAAEAAVQQLRIQPSKRRK
RUO
PCLO
Wheat Germ (in vitro)
GST
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit