Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PD-1 His-tag, Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

HEK293 cells Recombinant Protein

$1199.00 - $3430.00

Specifications

Recombinant Yes
Name SARS-CoV-2 Spike Protein (S-RBD) (aa330-524), His Tag
For Use With (Application) Control, ELISA, Western Blot
Conjugate Unlabeled
Format Lyophilized
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87687 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87687
1 mg
Each for $3,430.00
Add to cart
 
PIRP87686 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87686
200 µg
Each for $1,199.00
Add to cart
 
Description

Description

Cell-mediated immune responses are initiated by T lymphocytes that are themselves stimulated by cognate peptides bound to MHC molecules on antig en-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as PD-1 (also known as PDCD-1) that mediate inhibitory signals from the APC. PD-1 can bind two different but related ligands, PDL-1 and PDL-2. Upon binding to either of these ligands, signals generated by PD-1 inhibit the activation of the immune response in the absence of danger signals such as LPS or other molecules associated with bacteria or other pathogens. Evidence for this is seen in PD1-null mice who exhibit hyperactivated immune systems and autoimmune diseases. Despite its predicted molecular weight, PD-1 often migrates at higher molecular weight in SDS-PAGE.
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Lyophilized
Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
RUO
Virus
SARS-CoV-2 Spike Protein (S-RBD) (aa330-524), His Tag
Unlabeled
PBS with no preservative
HEK293 cells
-20°Cor to +80°C if preferred
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit