Learn More
Raybiotech IncSupplier Diversity Partner Human PD-ECGF (Thymidine Phosphorylase) ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHTYMP1
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.4 ng/mL. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human PD-ECGF. Other species not determined.
Catalog No. 50-194-9832