Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PDCD11 Partial ORF (AAH49838, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name PDCD11 (Human) Recombinant Protein (Q01)
Recombinant Yes
For Use With (Application) Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Molecular Weight 37.84kDa
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-956-153
View Documents
Abnova™
H00022984Q01L
25 ug
Each for $709.50
Add to cart
 
89-956-152
View Documents
Abnova™
H00022984Q01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

PDCD11 is a NF-kappa-B (NFKB1; 164011)-binding protein that colocalizes with U3 RNA (MIM 180710) in the nucleolus and is required for rRNA maturation and generation of 18S rRNA (Sweet et al., 2003 [PubMed 14624448]; Sweet et al., 2008 [PubMed 17654514]).[supplied by OMIM]

Sequence: MANLEESFPRGGTRKIHKPEKAFQQSVEQDNLFDISTEEGSTKRKKSQKGPAKTKKLKIEKRESSKSAREKFEILSVESLCEGMRILGCVKEVNELELVISLPNGLQGFV
Specifications

Specifications

PDCD11 (Human) Recombinant Protein (Q01)
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
37.84kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit