Learn More
Abnova™ Human PDGFA/PDGFB (heterodimer) Recombinant Protein
Used for Func, SDS-PAGE
Supplier: Abnova™ P4800
Description
The protein encoded by this gene is a member of the platelet-derived growth factor family. The four members of this family are mitogenic factors for cells of mesenchymal origin and are characterized by a motif of eight cysteines. This gene product can exist either as a homodimer (PDGF-BB) or as a heterodimer with the platelet-derived growth factor alpha polypeptide (PDGF-AB), where the dimers are connected by disulfide bonds. Mutations in this gene are associated with meningioma. Reciprocal translocations between chromosomes 22 and 7, at sites where this gene and that for COL1A1 are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans resulting from unregulated expression of growth factor. Two alternatively spliced transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq]
Sequence: PDGFA: MSIEEAVPAVCKTRTVIYEIPRSQVDPTSANFLIWPPCVEVKRCTGCCNTSSVKCQPSRVHHRSVKVAKVEYVRKKPKLKEVQVRLEEHLECACATTSLNPDYREEDTGRPRESGKKRKRKRLKPTSpecifications
P04085 (Gene ID : 5154);P01127 (Gene ID : 5155) | |
Lyophilized | |
13.3kDa | |
Escherichia coli expression system | |
Alpha chain: MSIEEAVPAVCKTRTVIYEIPRSQVDPTSANFLIWPPCVEVKRCTGCCNTSSVKCQPSRVHHRSVKVAKVEYVRKKPKLKEVQVRLEEHLECACATTSLNPDYREEDTGRPRESGKKRKRKRLKPT Beta chain: MSLGSLTIAEPAMIAECKTRTEVFEISRRLIDRTNANFLVWPPCVEVQRCSGCCNNRNVQCRPTQVQLRPVQVRKIEIVRKKPIFKKATVTLEDHLACKCETVAAARPVT | |
RUO | |
PDGF-A/PDGF1 | |
PDGFA | |
E. coli | |
None | |
Lyophilized |
Functional Study, SDS-PAGE | |
5154/5155 | |
PDGFA/PDGFB (Human) Recombinant Protein | |
10 ug | |
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
<0.1 EU/μg | |
PDGFA | |
The activity is determined by the dose-dependent proliferation of mouse 3T3 indicator cells. The expected ED50 for this effect is 1.4-2.1ng/mL. | |
Recombinant | |
Escherichia coli expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.