Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PDIA6 Full-length ORF (AAH01312, 1 a.a. - 440 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00010130P01S

 View more versions of this product

Catalog No. 89-966-263


Add to cart

Description

Description

Protein disulfide isomerases (EC 5.3.4.1), such as PDIA6, are endoplasmic reticulum (ER) resident proteins that catalyze formation, reduction, and isomerization of disulfide bonds in proteins and are thought to play a role in folding of disulfide-bonded proteins (Hayano and Kikuchi, 1995 [PubMed 7590364]).[supplied by OMIM]

Sequence: MALLVLGLVSCTFFLAVNGLYSSSDDVIELTPSNFNREVIQSDSLWLVEFYAPWCGHCQRLTPEWKKAATALKDVVKVGAVDADKHHSLGGQYGVQGFPTIKIFGSNKNRPEDYQGGRTGEAIVDAALSALRQLVKDRLGGRSGGYSSGKQGRSDSSSKKDVIELTDDSFDRNVLDSEDVWMVEFYAPWCGHCKNLEPEWAAAASEVKEQTKGKVKLAAVDATVNQVLASRYGIRGFPTIKIFQKGESPVDYDGGRTRSDIVSRALDLFSDNAPPPELLEIINEDIAKRTCEEHQLCVVAVLPHILDTGAAGRNSYLEVLLKLADKYKKKMWGWLWTEAGAQSELETALGIGGFGYPAMAAINARKMKFALLKGSFSEQGINEFLRELSFGRGSTAPVGGGAFPTIVEREPWDGRDGELPVEGDIDLSDVELDDLGKDEL
Specifications

Specifications

AAH01312
Liquid
10130
PDIA6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MALLVLGLVSCTFFLAVNGLYSSSDDVIELTPSNFNREVIQSDSLWLVEFYAPWCGHCQRLTPEWKKAATALKDVVKVGAVDADKHHSLGGQYGVQGFPTIKIFGSNKNRPEDYQGGRTGEAIVDAALSALRQLVKDRLGGRSGGYSSGKQGRSDSSSKKDVIELTDDSFDRNVLDSEDVWMVEFYAPWCGHCKNLEPEWAAAASEVKEQTKGKVKLAAVDATVNQVLASRYGIRGFPTIKIFQKGESPVDYDGGRTRSDIVSRALDLFSDNAPPPELLEIINEDIAKRTCEEHQLCVVAVLPHILDTGAAGRNSYLEVLLKLADKYKKKMWGWLWTEAGAQSELETALGIGGFGYPAMAAINARKMKFALLKGSFSEQGINEFLRELSFGRGSTAPVGGGAFPTIVEREPWDGRDGELPVEGDIDLSDVELDDLGKDEL
RUO
PDIA6
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
74.14kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ERP5|P5|TXNDC7
PDIA6
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit