Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PELI3 Full-length ORF (AAH21256.1, 1 a.a. - 331 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00246330P01L

 View more versions of this product

Catalog No. 89-956-408


Add to cart

Description

Description

Toll-like receptors (TLRs; see MIM 603030) and IL1R (IL1R1; MIM 147810) are part of the innate immune response aimed at mobilizing defense mechanisms in response in infection or injury. Pellino proteins, such as PELI3, are intermediate components in the signaling cascades initiated by TLRs and IL1R (Jensen and Whitehead, 2003 [PubMed 12874243]).[supplied by OMIM]

Sequence: MVLEGNPEVGSPRTSDLQHRGNKGSCVLSSPGEDAQPGEEPIKYGELIVLGCCEEGGEETEAQRGEVTGPRAHSCYNGCLASGDKGRRRSRLALSRRSHANGVKPDVMHHISTPLVSKALSNRGQHSISYTLSRSHSVIVEYTHDSDTDMFQIGRSTENMIDFVVTDTSPGGGAAEGPSAQSTISRYACRILCDRRPPYTARIYAAGFDASSNIFLGERAAKWRTPDGLMDGLTTNGVLVMHPAGGFSEDSAPGVWREISVCGNVYTLRDSRSAQQRGKLVGGPLHSPPLSLPGPHKLPIPLPKPGDDLQSFCPREPQQAPYQGIPGPGGS
Specifications

Specifications

AAH21256.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
61.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC35521
PELI3
Yes
wheat germ expression system
Liquid
246330
PELI3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVLEGNPEVGSPRTSDLQHRGNKGSCVLSSPGEDAQPGEEPIKYGELIVLGCCEEGGEETEAQRGEVTGPRAHSCYNGCLASGDKGRRRSRLALSRRSHANGVKPDVMHHISTPLVSKALSNRGQHSISYTLSRSHSVIVEYTHDSDTDMFQIGRSTENMIDFVVTDTSPGGGAAEGPSAQSTISRYACRILCDRRPPYTARIYAAGFDASSNIFLGERAAKWRTPDGLMDGLTTNGVLVMHPAGGFSEDSAPGVWREISVCGNVYTLRDSRSAQQRGKLVGGPLHSPPLSLPGPHKLPIPLPKPGDDLQSFCPREPQQAPYQGIPGPGGS
RUO
PELI3
Wheat Germ (in vitro)
GST
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit