Learn More
Abnova™ Human PEO1 Partial ORF (NP_068602.2, 591 a.a. - 684 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_068602.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 56652 |
Molecular Weight (g/mol) | 36.08kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-956-419
|
Abnova™
H00056652Q01L |
25 ug |
Each for $769.00
|
|
|||||
89-956-418
|
Abnova™
H00056652Q01S |
10 ug |
Each for $499.00
|
|
|||||
Description
This gene encodes a hexameric DNA helicase which unwinds short stretches of double-stranded DNA in the 5' to 3' direction and, along with mitochondrial single-stranded DNA binding protein and mtDNA polymerase gamma, is thought to play a key role in mtDNA replication. The protein localizes to the mitochondrial matrix and mitochondrial nucleoids. Mutations in this gene cause infantile onset spinocerebellar ataxia (IOSCA) and progressive external ophthalmoplegia (PEO) and are also associated with several mitochondrial depletion syndromes. Alternative splicing results in multiple transcript variants encoding distinct isoforms
Sequence: DRKLVTGPGKRYLQVSKNRFDGDVGVFPLEFNKNSLTFSIPPKNKARLKKIKDDTGPVAKKPSSGKKGATTQNSEICSGQAPTPDQPDTSKRSKSpecifications
NP_068602.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.08kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATXN8/FLJ21832/IOSCA/PEO/PEO1/PEOA3/SANDO/SCA8/TWINL | |
C10orf2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
56652 | |
PEO1 (Human) Recombinant Protein (Q01) | |
DRKLVTGPGKRYLQVSKNRFDGDVGVFPLEFNKNSLTFSIPPKNKARLKKIKDDTGPVAKKPSSGKKGATTQNSEICSGQAPTPDQPDTSKRSK | |
RUO | |
C10orf2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.