Learn More
Novus Biologicals™ Human Peptide YY ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP262166
Catalog No. NBP262166
Add to cart
Description
Description
The Human Peptide YY ELISA Kit (Colorimetric) measures human Peptide YY in human serum, plasma, cell culture supernatant, tissue homogenate.
Specifications
Specifications
ELISA Kit (Colorimetric)
ELISA
Store at 4°C
Human
Cell Culture Supernatant, Human Serum, Plasma, Tissue Homogenates
PYY
31.25-2000 pg/mL
5697
Peptide YY
1 Kit
18.75 pg/mL