Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PES1 Partial ORF (NP_055118.1, 3 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00023481Q01L

 View more versions of this product

Catalog No. 89-956-441


Add to cart

Description

Description

This gene encodes a protein that is abnormally elevated in malignant tumors of astrocytic origin. It is a strongly conserved gene containing a BRCT domain that is essential for the activity of this gene product. The gene plays a crucial role in cell proliferation and may be necessary for oncogenic transformation and tumor progression. [provided by RefSeq]

Sequence: GLEKKKYERGSATNYITRNKARKKLQLSLADFRRLCILKGIYPHEPKHKKKVNKGSTAARTFYLIKDIRFLLHEPIVNKFREYKVFVRKLRKAYGKSE
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GLEKKKYERGSATNYITRNKARKKLQLSLADFRRLCILKGIYPHEPKHKKKVNKGSTAARTFYLIKDIRFLLHEPIVNKFREYKVFVRKLRKAYGKSE
RUO
PES1
Wheat Germ (in vitro)
GST
NP_055118.1
Liquid
23481
PES1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PES
PES1
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit