Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PHEMX Partial ORF (NP_005696, 194 a.a. - 290 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00010077Q01S

 View more versions of this product

Catalog No. 89-956-658


Add to cart

Description

Description

This gene, which is a member of the tetraspanin superfamily, is one of several tumor-suppressing subtransferable fragments located in the imprinted gene domain of chromosome 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian and breast cancers. This gene is located among several imprinted genes; however, this gene, as well as the tumor-suppressing subchromosomal transferable fragment 4, escapes imprinting. This gene may play a role in malignancies and diseases that involve this region, and it is also involved in hematopoietic cell function. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq]

Sequence: RCGCSLDRKGKYTLTPRACGRQPQEPSLLRCSQGGPTHCLHSEAVAIGPRGCSGSLRWLQESDAAPLPLSCHLAAHRALQGRSRGGLSGCPERGLSD
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RCGCSLDRKGKYTLTPRACGRQPQEPSLLRCSQGGPTHCLHSEAVAIGPRGCSGSLRWLQESDAAPLPLSCHLAAHRALQGRSRGGLSGCPERGLSD
RUO
TSPAN32
Wheat Germ (in vitro)
GST
NP_005696
Liquid
10077
PHEMX (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ17158/FLJ97586/MGC22455/PHEMX/PHMX/TSSC6
TSPAN32
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit