Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PHF7 Full-length ORF (NP_057567.3, 1 a.a. - 381 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name PHF7 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 70.2kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-956-699
View Documents
Abnova™
H00051533P01L
25 ug
Each for $709.50
Add to cart
 
89-956-698
View Documents
Abnova™
H00051533P01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

Spermatogenesis is a complex process regulated by extracellular and intracellular factors as well as cellular interactions among interstitial cells of the testis, Sertoli cells, and germ cells. In the testis, this gene is expressed in Sertoli cells but not germ cells. However, this gene is not expressed in a patient who exhibited spermatogenic arrest at the spermatocyte stage. Spermatogenic arrest is an interruption of germ cell differentiation that may result in oligospermia or azoospermia. The proteins encoded by this gene contain plant homeodomain (PHD) finger domains, also known as leukemia associated protein (LAP) domains, believed to be involved in transcriptional regulation. Thus this protein, which localizes to the nucleus of transfected cells, has been implicated in the transcriptional regulation of spermatogenesis. Two protein isoforms are encoded by transcript variants of this gene. [provided by RefSeq]

Sequence: MKTVKEKKECQRLRKSAKTRRVTQRKPSSGPVCWLCLREPGDPEKLGEFLQKDNISVHYFCLILSSKLPQRGQSNRGFHGFLPEDIKKEAARASRKICFVCKKKGAAINCQKDQCLRNFHLPCGQERGCLSQFFGEYKSFCDKHRPTQNIQHGHVGEESCILCCEDLSQQSVENIQSPCCSQAIYHRKCIQKYAHTSAKHFFKCPQCNNRKEFPQEMLRMGIHIPDRDAAWELEPGAFSDLYQRYQHCDAPICLYEQGRDSFEDEGRWCLILCATCGSHGTHRDCSSLRSNSKKWECEECSPAAATDYIPENSGDIPCCSSTFHPEEHFCRDNTLEENPGLSWTDWPEPSLLEKPESSRGRRSYSWRSKGVRITNSCKKSK
Specifications

Specifications

PHF7 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
70.2kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit