Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Phospho-CD117/c-kit DuoSet IC ELISA

Human Phospho-CD117/c-kit DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

$545.20 - $2117.00

Specifications

Product Type ELISA Development Kit for Phosphorylated Intracellular Proteins
Quantity 1 Kit
Species Human
Conjugate Biotin
For Use With (Application) ELISA
View More Specs

Products 3
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
DYC3527E
View Documents
R&D Systems™
DYC3527E
15 Plates
Each for $2,117.00
Add to Cart
 
DYC35275
View Documents
R&D Systems™
DYC35275
5 Plates
Each for $997.60
Add to Cart
 
DYC35272
View Documents
R&D Systems™
DYC35272
2 Plates
Each for $545.20
Add to Cart
 
Description

Description

ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. This kit is designed for the detection and enumeration of IFN-γ/IL-2. Complete ELISpot kits are ready-to-run and require no assay development or refinement.

The Human Phospho-CD117/c-kit DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human CD117/c-kit.
Specifications

Specifications

ELISA Development Kit for Phosphorylated Intracellular Proteins
Human
ELISA
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
1 Kit
Biotin
3815
SDS
Documents

Documents

Product Certifications
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.