Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Phospho-Chk2 (T68) Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

$214.00 - $526.00

Specifications

Antigen Chk2
Clone 1238H
Dilution Western Blot 2 ug/mL, Immunocytochemistry 2-25 ug/mL
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1626SP
GREENER_CHOICE
View Documents
R&D Systems
MAB1626SP
25 μg
Each for $214.00
Only null left
Add to Cart
 
MAB1626100
GREENER_CHOICE
View Documents
R&D Systems
MAB1626100
100 μg
Each for $526.00
Only null left
Add to Cart
 
Description

Description

Chk2 Monoclonal specifically detects Chk2 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications

Specifications

Chk2
Western Blot 2 ug/mL, Immunocytochemistry 2-25 ug/mL
Monoclonal
Purified
RUO
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
CDS1, CHEK2, CHK2 checkpoint homolog (S. pombe), EC 2.7.11, EC 2.7.11.1, HuCds1, LFS2, PP1425, Rad53, S.pombe) homolog
CHEK2
IgG
Protein A or G purified from cell culture supernatant
Detects human Chk2 when phosphorylated at T68 in direct ELISAs and Western blots.
1238H
Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human
O96017
11200
E.coli-derived recombinant human Phospho-Chk2 (T68) Accession # O96017
Primary
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.