Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PIGS Full-length ORF (NP_149975.1, 1 a.a. - 555 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00094005P01S

 View more versions of this product

Catalog No. 89-956-923


Add to cart

Description

Description

This gene encodes a protein that is involved in GPI-anchor biosynthesis. The glycosylphosphatidylinositol (GPI) anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. This gene encodes an essential component of the multisubunit enzyme, GPI transamidase. GPI transamidase mediates GPI anchoring in the endoplasmic reticulum, by catalyzing the transfer of fully assembled GPI units to proteins. [provided by RefSeq]

Sequence: MAAAGAAATHLEVARGKRAALFFAAVAIVLGLPLWWKTTETYRASLPYSQISGLNALQLRLMVPVTVVFTRESVPLDDQEKLPFTVVHEREIPLKYKMKIKCRFQKAYRRALDHEEEALSSGSVQEAEAMLDEPQEQAEGSLTVYVISEHSSLLPQDMMSYIGPKRTAVVRGIMHREAFNIIGRRIVQVAQAMSLTEDVLAAALADHLPEDKWSAEKRRPLKSSLGYEITFSLLNPDPKSHDVYWDIEGAVRRYVQPFLNALGAAGNFSVDSQILYYAMLGVNPRFDSASSSYYLDMHSLPHVINPVESRLGSSAASLYPVLNFLLYVPELAHSPLYIQDKDGAPVATNAFHSPRWGGIMVYNVDSKTYNASVLPVRVEVDMVRVMEVFLAQLRLLFGIAQPQLPPKCLLSGPTSEGLMTWELDRLLWARSVENLATATTTLTSLAQLLGKISNIVIKDDVASEVYKAVAAVQKSAEELASGHLASAFVASQEAVTSSELAFFDPSLLHLLYFPDDQKFAIYIPLFLPMAVPILLSLVKIFLETRKSWRKPEKTD
Specifications

Specifications

NP_149975.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
88.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686K20216|FLJ45226
PIGS
Yes
wheat germ expression system
Liquid
94005
PIGS (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAAGAAATHLEVARGKRAALFFAAVAIVLGLPLWWKTTETYRASLPYSQISGLNALQLRLMVPVTVVFTRESVPLDDQEKLPFTVVHEREIPLKYKMKIKCRFQKAYRRALDHEEEALSSGSVQEAEAMLDEPQEQAEGSLTVYVISEHSSLLPQDMMSYIGPKRTAVVRGIMHREAFNIIGRRIVQVAQAMSLTEDVLAAALADHLPEDKWSAEKRRPLKSSLGYEITFSLLNPDPKSHDVYWDIEGAVRRYVQPFLNALGAAGNFSVDSQILYYAMLGVNPRFDSASSSYYLDMHSLPHVINPVESRLGSSAASLYPVLNFLLYVPELAHSPLYIQDKDGAPVATNAFHSPRWGGIMVYNVDSKTYNASVLPVRVEVDMVRVMEVFLAQLRLLFGIAQPQLPPKCLLSGPTSEGLMTWELDRLLWARSVENLATATTTLTSLAQLLGKISNIVIKDDVASEVYKAVAAVQKSAEELASGHLASAFVASQEAVTSSELAFFDPSLLHLLYFPDDQKFAIYIPLFLPMAVPILLSLVKIFLETRKSWRKPEKTD
RUO
PIGS
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit