Learn More
Abnova™ Human PK1 (NM_004938, 1 a.a. - 363 a.a.) Full-length Recombinant Protein with GST-His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4674
Catalog No. 89-941-412
Add to cart
Description
Description
Death-associated protein kinase 1 is a positive mediator of gamma-interferon induced programmed cell death. DAPK1 encodes a structurally unique 160-kD calmodulin dependent serine-threonine kinase that carries 8 ankyrin repeats and 2 putative P-loop consensus sites. It is a tumor suppressor candidate. [provided by RefSeq]
Specifications
Specifications
NM_004938
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
71.227kDa
Insect cell (Sf9) expression system
2 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing
DAPK|DKFZp781I035
DAPK1
85pmol/μg x min
Yes
0.680 ug/uL
Liquid
1612
DAPK1 (Human) Recombinant Protein
One-step affinity purification using GSH-agarose
100 ug
RUO
DAPK1
Insect cell (Sf9) expression system
Insect
GST-His