Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PKD2L1 Full-length ORF (NP_057196.2, 1 a.a. - 805 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009033P02S

 View more versions of this product

Catalog No. 89-957-097


Add to cart

Description

Description

This gene encodes a member of the polycystin protein family. The encoded protein contains multiple transmembrane domains, and cytoplasmic N- and C-termini. The protein may be an integral membrane protein involved in cell-cell/matrix interactions. This protein functions as a calcium-regulated nonselective cation channel. Alternative splice variants have been described but their full length sequences have not been determined. [provided by RefSeq]

Sequence: MNAVGSPEGQELQKLGSGAWDNPAYSGPPSPHGTLRVCTISSTGPLQPQPKKPEDEPQETAYRTQVSSCCLHICQGIRGLWGTTLTENTAENRELYIKTTLRELLVYIVFLVDICLLTYGMTSSSAYYYTKVMSELFLHTPSDTGVSFQAISSMADFWDFAQGPLLDSLYWTKWYNNQSLGHGSHSFIYYENMLLGVPRLRQLKVRNDSCVVHEDFREDILSCYDVYSPDKEEQLPFGPFNGTAWTYHSQDELGGFSHWGRLTSYSGGGYYLDLPGSRQGSAEALRALQEGLWLDRGTRVVFIDFSVYNANINLFCVLRLVVEFPATGGAIPSWQIRTVKLIRYVSNWDFFIVGCEVIFCVFIFYYVVEEILELHIHRLRYLSSIWNILDLVVILLSIVAVGFHIFRTLEVNRLMGKLLQQPNTYADFEFLAFWQTQYNNMNAVNLFFAWIKIFKYISFNKTMTQLSSTLARCAKDILGFAVMFFIVFFAYAQLGYLLFGTQVENFSTFIKCIFTQFRIILGDFDYNAIDNANRILGPAYFVTYVFFVFFVLLNMFLAIINDTYSEVKEELAGQKDELQLSDLLKQGYNKTLLRLRLRKERVSDVQKVLQGGEQEIQFEDFTNTLRELGHAEHEITELTATFTKFDRDGNRILDEKEQEKMRQDLEEERVALNTEIEKLGRSIVSSPQGKSGPEAARAGGWVSGEEFYMLTRRVLQLETVLEGVVSQIDAVGSKLKMLERKGWLAPSPGVKEQAIWKHPQPAPAVTPDPWGVQGGQESEVPYKREEEALEERRLSRGEIPTLQRS
Specifications

Specifications

NP_057196.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
118.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PCL|PKD2L|PKDL
PKD2L1
Yes
wheat germ expression system
Liquid
9033
PKD2L1 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNAVGSPEGQELQKLGSGAWDNPAYSGPPSPHGTLRVCTISSTGPLQPQPKKPEDEPQETAYRTQVSSCCLHICQGIRGLWGTTLTENTAENRELYIKTTLRELLVYIVFLVDICLLTYGMTSSSAYYYTKVMSELFLHTPSDTGVSFQAISSMADFWDFAQGPLLDSLYWTKWYNNQSLGHGSHSFIYYENMLLGVPRLRQLKVRNDSCVVHEDFREDILSCYDVYSPDKEEQLPFGPFNGTAWTYHSQDELGGFSHWGRLTSYSGGGYYLDLPGSRQGSAEALRALQEGLWLDRGTRVVFIDFSVYNANINLFCVLRLVVEFPATGGAIPSWQIRTVKLIRYVSNWDFFIVGCEVIFCVFIFYYVVEEILELHIHRLRYLSSIWNILDLVVILLSIVAVGFHIFRTLEVNRLMGKLLQQPNTYADFEFLAFWQTQYNNMNAVNLFFAWIKIFKYISFNKTMTQLSSTLARCAKDILGFAVMFFIVFFAYAQLGYLLFGTQVENFSTFIKCIFTQFRIILGDFDYNAIDNANRILGPAYFVTYVFFVFFVLLNMFLAIINDTYSEVKEELAGQKDELQLSDLLKQGYNKTLLRLRLRKERVSDVQKVLQGGEQEIQFEDFTNTLRELGHAEHEITELTATFTKFDRDGNRILDEKEQEKMRQDLEEERVALNTEIEKLGRSIVSSPQGKSGPEAARAGGWVSGEEFYMLTRRVLQLETVLEGVVSQIDAVGSKLKMLERKGWLAPSPGVKEQAIWKHPQPAPAVTPDPWGVQGGQESEVPYKREEEALEERRLSRGEIPTLQRS
RUO
PKD2L1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit