Learn More
Abnova™ Human PKN2 (NP_006247.1, 1 a.a. - 984 a.a.) Full-length Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5624
Catalog No. 89-957-131
Add to cart
Specifications
Specifications
NP_006247.1
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
139kDa
Baculovirus infected insect cell (Sf21) expression system
91% by SDS-PAGE/CBB staining
5 ug
RUO
PKN2
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: GS peptide. ATP: 100 uM.
Yes
Baculovirus infected insect cell (Sf21) expression system
Liquid
5586
PKN2 (Human) Recombinant Protein
Glutathione sepharose chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC150606|MGC71074|PAK2|PRK2|PRKCL2|PRO2042|Pak-2
PKN2
Insect
GST