Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human POU4F1 Full-length ORF (AAI48793.1, 1 a.a. - 419 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00005457P01

Catalog No. 89-957-696


Add to cart

Description

Description

BRN3A (POU4F1) is a class IV POU domain-containing transcription factor highly expressed in the developing sensory nervous system and in cells of the B- and T-lymphocytic lineages (Gerrero et al., 1993 [PubMed 8248179]).[supplied by OMIM]

Sequence: MMSMNSKQPHFAMHPTLPEHKYPSLHSSSEAIRRACLPTPPLQSNLFASLDETLLARAEALAAVDIAVSQGKSHPFKPDATYHTMNSVPCTSTSTVPLAHHHHHHHHHQALEPGDLLDHISSPSLALMAGAGGAGAAAGGGGAHDGPGGGGGPGGGGGPGGGPGGGGGGGPGGGGGGPGGGLLGGSAHPHPHMHSLGHLSHPAAAAAMNMPSGLPHPGLVAAAAHHGAAAAAAAAAAGQVAAASAAAAVVGAAGLASICDSDTDPRELEAFAERFKQRRIKLGVTQADVGSALANLKIPGVGSLSQSTICRFESLTLSHNNMIALKPILQAWLEEAEGAQREKMNKPELFNGGEKKRKRTSIAAPEKRSLEAYFAVQPRPSSEKIAAIAEKLDLKKNVVRVWFCNQRQKQKRMKFSATY
Specifications

Specifications

AAI48793.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
73.04kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BRN3A|FLJ13449|Oct-T1|RDC-1
POU4F1
Yes
wheat germ expression system
Liquid
5457
POU4F1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MMSMNSKQPHFAMHPTLPEHKYPSLHSSSEAIRRACLPTPPLQSNLFASLDETLLARAEALAAVDIAVSQGKSHPFKPDATYHTMNSVPCTSTSTVPLAHHHHHHHHHQALEPGDLLDHISSPSLALMAGAGGAGAAAGGGGAHDGPGGGGGPGGGGGPGGGPGGGGGGGPGGGGGGPGGGLLGGSAHPHPHMHSLGHLSHPAAAAAMNMPSGLPHPGLVAAAAHHGAAAAAAAAAAGQVAAASAAAAVVGAAGLASICDSDTDPRELEAFAERFKQRRIKLGVTQADVGSALANLKIPGVGSLSQSTICRFESLTLSHNNMIALKPILQAWLEEAEGAQREKMNKPELFNGGEKKRKRTSIAAPEKRSLEAYFAVQPRPSSEKIAAIAEKLDLKKNVVRVWFCNQRQKQKRMKFSATY
RUO
POU4F1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit