Learn More
Abnova™ Human POU6F2 Partial ORF (NP_009183, 2 a.a. - 87 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00011281Q01L
Description
POU6F2 is a member of a gene family characterized by the presence of a bipartite DNA-binding domain, consisting of a POU-specific domain and a POU heterodomain, separated by a variable polylinker. POU domain family members are transcriptional regulators, many of which show highly restricted patterns of expression and are known to control cell type-specific differentiation pathways (see review by Phillips and Luisi, 2000 [PubMed 11183772]).[supplied by OMIM]
Sequence: IAGQVSKPLLSVRSEMNAELRGEDKAATSDSELNEPLLAPVESNDSEDTPSKLFGARGNPALSDPGTPDQHQASQTHPPFPVGPQPSpecifications
NP_009183 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.2kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
IAGQVSKPLLSVRSEMNAELRGEDKAATSDSELNEPLLAPVESNDSEDTPSKLFGARGNPALSDPGTPDQHQASQTHPPFPVGPQP | |
RUO | |
POU6F2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
11281 | |
POU6F2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RPF-1/WT5/WTSL | |
POU6F2 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.