Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Abnova™ Human PPP1R9A Partial ORF (XP_371933, 1090 a.a. - 1188 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00055607Q01L

 View more versions of this product

Catalog No. 89-957-904



Description

Description

Sequence: PCSTAQTSTRSPCMPFSWFNDSRKGSYSFRNLPAPTSSLQPSPETLISDKKGSKNFTFNDDFSPSSTSSADLSGLGAEPKTPGLSQSLALSSDESLDMI
Specifications

Specifications

XP_371933
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PCSTAQTSTRSPCMPFSWFNDSRKGSYSFRNLPAPTSSLQPSPETLISDKKGSKNFTFNDDFSPSSTSSADLSGLGAEPKTPGLSQSLALSSDESLDMI
RUO
PPP1R9A
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
55607
PPP1R9A (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ20068|KIAA1222|NRB1|NRBI|Neurabin-I
PPP1R9A
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit