Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PRICKLE1 Partial ORF (NP_694571, 684 a.a. - 756 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00144165Q01S

 View more versions of this product

Catalog No. 89-958-092


Add to cart

Description

Description

This gene encodes a nuclear receptor that may be a negative regulator of the Wnt/beta-catenin signaling pathway. The encoded protein localizes to the nuclear membrane and has been implicated in the nuclear trafficking of the transcription repressors REST/NRSF and REST4. Mutations in this gene have been linked to progressive myoclonus epilepsy. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 3. [provided by RefSeq]

Sequence: DNALNLVTERKYSPKDRLRLYTPDNYEKFIQNKSAREIQAYIQNADLYGQYAHATPDYGLQNPGMNRFLGLYG
Specifications

Specifications

NP_694571
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.77kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DNALNLVTERKYSPKDRLRLYTPDNYEKFIQNKSAREIQAYIQNADLYGQYAHATPDYGLQNPGMNRFLGLYG
RUO
PRICKLE1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
144165
PRICKLE1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EPM1B|FLJ31627|FLJ31937|MGC138902|MGC138903|RILP
PRICKLE1
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit