Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Primate IL-17/IL-17A Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 11 publications
Supplier: R&D Systems BAF317
This item is not returnable.
View return policy
Description
IL-17/IL-17A Polyclonal specifically detects IL-17/IL-17A in Human, Primate samples. It is validated for Immunohistochemistry, ELISA Detection (Matched Antibody Pair).Specifications
IL-17/IL-17A | |
Polyclonal | |
Biotin | |
Q16552 | |
IL17A | |
E. coli-derived recombinant human IL-17 Ile20-Ala155 Accession # Q16552 | |
50 μg | |
Primary | |
Detects human and primate IL-17 in direct ELISAs. In sandwich immunoassays, approximately 22% cross-reactivity with rhIL-17A/F heterodimer is observed, and less than 0.05% cross-reactivity with recombinant mouse (rm) IL-17, recombinant human (rh) IL-10, rhIL-12, rhIL-16, rhIFN-gamma , rhIL-17B, rhIL-17C, rhIL-17D, rhIL-17E and rhIL-17F is observed. | |
Human, Primate | |
IgG |
Immunohistochemistry, ELISA | |
LYOPH | |
Immunohistochemistry 0.3-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
Goat | |
Affinity Purified | |
RUO | |
3605 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction