Learn More
Abnova Human PRKCZ (Z15108, 6 a.a. - 584 a.a.) Partial Recombinant Protein with GST-His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer: Abnova P4764
Description
Protein kinase C (PKC) zeta is a member of the PKC family of serine/threonine kinases which are involved in a variety of cellular processes such as proliferation, differentiation and secretion. Unlike the classical PKC isoenzymes which are calcium-dependent, PKC zeta exhibits a kinase activity which is independent of calcium and diacylglycerol but not of phosphatidylserine. Furthermore, it is insensitive to typical PKC inhibitors and cannot be activated by phorbol ester. Unlike the classical PKC isoenzymes, it has only a single zinc finger module. These structural and biochemical properties indicate that the zeta subspecies is related to, but distinct from other isoenzymes of PKC. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Specifications
Z15108 | |
0.181 ug/uL | |
Liquid | |
5590 | |
PRKCZ (Human) Recombinant Protein | |
One-step affinity purification using GSH agarose | |
100 ug | |
RUO | |
PRKCZ | |
83pmol/μg x min | |
Yes |
Insect cell (Sf9) expression system | |
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis | |
Liquid | |
96.654kDa | |
Insect cell (Sf9) expression system | |
2 ug/lane SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing | |
PKC-ZETA|PKC2 | |
PRKCZ | |
Insect | |
GST-His |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.