Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Abnova™ Human PRKRA Partial ORF (NP_003681, 101 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00008575Q01S

 View more versions of this product

Catalog No. 89-958-223



Description

Description

This gene encodes a protein kinase activated by double-stranded RNA which mediates the effects of interferon in response to viral infection. Mutations in this gene have been associated with dystonia. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: ANASICFAVPDPLMPDPSKQPKNQLNPIGSLQELAIHHGWRLPEYTLSQEGGPAHKREYTTICRLESFMETGKGASKKQAKRNAAEKFLAKFSNISPENH
Specifications

Specifications

NP_003681
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ANASICFAVPDPLMPDPSKQPKNQLNPIGSLQELAIHHGWRLPEYTLSQEGGPAHKREYTTICRLESFMETGKGASKKQAKRNAAEKFLAKFSNISPENH
RUO
PRKRA
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
8575
PRKRA (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DYT16|HSD14|PACT|RAX
PRKRA
Yes
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit