Learn More
Novus Biologicals™ Human Progranulin/PGRN ELISA Kit (Colorimetric)
This kit recognizes Human PGRN in samples.
Manufacturer:  Novus Biologicals™ NBP280229
Catalog No. NB013300
Add to cart
Description
Description
No significant cross-reactivity or interference between Human PGRN and analogues was observed.
Specifications
Specifications
62.5 to 4000 pg/mL
ELISA Kit (Colorimetric)
This kit recognizes Human PGRN in samples. No significant cross-reactivity or interference between Human PGRN and analogues was observed.
ELISA
GRN
1 Kit
CLN11, GEP, GP88, granulin precursor, GRN, PCDGF, PEPI, PGRN
37.5 pg/mL
Serum, Plasma, Other Biological Fluids
Progranulin/PGRN
2896
Human
Storage of components varies. See protocol for specific instructions.
Progranulin/PGRN