Learn More
ACROBiosystems ACROBiosystems Human Prolactin / PRL Protein, Mouse IgG2a Fc Tag, low endotoxin
Supplier:  ACROBiosystems PRNH5257100UG
Encompass_Preferred
Recombinant Protein;100UG;Human Prolactin, Mouse IgG2a Fc Tag, low endotoxin (PRN-H5257) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Cys 227 (Accession # Q5THQ0-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 49.8 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Prolactin, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. PRN-H5257) at 1 μg/mL (100 μL/well) can bind Human Prolactin R, Fc Tag (Cat. No. PRP-H5251) with a linear range of 2-39 ng/mL (QC tested).
Catalog No. 50-210-6718