Learn More
Abnova Human PRPH Full-length ORF (NP_006253.2, 1 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova H00005630P01S
Description
This gene encodes a cytoskeletal protein found in neurons of the peripheral nervous system. The encoded protein is a type III intermediate filament protein with homology to other cytoskeletal proteins such as desmin, and is a different protein that the peripherin found in photoreceptors. Mutations in this gene have been associated with susceptibility to amyotrophic lateral sclerosis. [provided by RefSeq]
Sequence: MSHHPSGLRAGFSSTSYRRTFGPPPSLSPGAFSYSSSSRFSSSRLLGSASPSSSVRLGSFRSPRAGAGALLRLPSERLDFSMAEALNQEFLATRSNEKQELQELNDRFANFIEKVRFLEQQNAALRGELSQARGQEPARADQLCQQELRELRRELELLGRERDRVQVERDGLAEDLAALKQRLEEETRKREDAEHNLVLFRKDVDDATLSRLELERKIESLMDEIEFLKKLHEEELRDLQVSVESQQVQQVEVEATVKPELTAALRDIRAQYESIAAKNLQEAEEWYKSKYADLSDAANRNHEALRQAKQEMNESRRQIQSLTCEVDGLRGTNEALLRQLRELEEQFALEAGGYQAGAARLEEELRQLKEEMARHLREYQELLNVKMALDIEIATYRKLLEGEESRISVPVHSFASLNIKTTVPEVEPPQDSHSRKTVLIKTIETRNGEVVTESQKEQRSELDKSSAHSYSpecifications
NP_006253.2 | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
80.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NEF4|PRPH1 | |
PRPH | |
Yes |
wheat germ expression system | |
Liquid | |
5630 | |
PRPH (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSHHPSGLRAGFSSTSYRRTFGPPPSLSPGAFSYSSSSRFSSSRLLGSASPSSSVRLGSFRSPRAGAGALLRLPSERLDFSMAEALNQEFLATRSNEKQELQELNDRFANFIEKVRFLEQQNAALRGELSQARGQEPARADQLCQQELRELRRELELLGRERDRVQVERDGLAEDLAALKQRLEEETRKREDAEHNLVLFRKDVDDATLSRLELERKIESLMDEIEFLKKLHEEELRDLQVSVESQQVQQVEVEATVKPELTAALRDIRAQYESIAAKNLQEAEEWYKSKYADLSDAANRNHEALRQAKQEMNESRRQIQSLTCEVDGLRGTNEALLRQLRELEEQFALEAGGYQAGAARLEEELRQLKEEMARHLREYQELLNVKMALDIEIATYRKLLEGEESRISVPVHSFASLNIKTTVPEVEPPQDSHSRKTVLIKTIETRNGEVVTESQKEQRSELDKSSAHSY | |
RUO | |
PRPH | |
Wheat Germ (in vitro) | |
GST |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.