Learn More
Abnova Human PRPSAP2 Full-length ORF (NP_002758.1, 1 a.a. - 369 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova H00005636P01S
Description
The enzyme phosphoribosylpyrophosphate synthetase (PRS) catalyzes the formation of phosphoribosylpyrophosphate which is a substrate for synthesis of purine and pyrimidine nucleotides, histidine, tryptophan and NAD. PRS exists as a complex with two catalytic subunits and two associated subunits. This gene encodes a non-catalytic associated subunit of PRS. [provided by RefSeq]
Sequence: MFCVTPPELETKMNITKGGLVLFSANSNSSCMELSKKIAERLGVEMGKVQVYQEPNRETRVQIQESVRGKDVFIIQTVSKDVNTTIMELLIMVYACKTSCAKSIIGVIPYFPYSKQCKMRKRGSIVSKLLASMMCKAGLTHLITMDLHQKEIQGFFNIPVDNLRASPFLLQYIQEEIPDYRNAVIVAKSPASAKRAQSFAERLRLGIAVIHGEAQDAESDLVDGRHSPPMVRSVAAIHPSLEIPMLIPKEKPPITVVGDVGGRIAIIVDDIIDDVDSFLAAAETLKERGAYKIFVMATHGLLSSDAPRRIEESAIDEVVVTNTIPHEVQKLQCPKIKTVDISMILSEAIRRIHNGESMSYLFRNIGLDDSpecifications
NP_002758.1 | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
67.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC117304|MGC126719|MGC126721|PAP41 | |
PRPSAP2 | |
Yes |
wheat germ expression system | |
Liquid | |
5636 | |
PRPSAP2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MFCVTPPELETKMNITKGGLVLFSANSNSSCMELSKKIAERLGVEMGKVQVYQEPNRETRVQIQESVRGKDVFIIQTVSKDVNTTIMELLIMVYACKTSCAKSIIGVIPYFPYSKQCKMRKRGSIVSKLLASMMCKAGLTHLITMDLHQKEIQGFFNIPVDNLRASPFLLQYIQEEIPDYRNAVIVAKSPASAKRAQSFAERLRLGIAVIHGEAQDAESDLVDGRHSPPMVRSVAAIHPSLEIPMLIPKEKPPITVVGDVGGRIAIIVDDIIDDVDSFLAAAETLKERGAYKIFVMATHGLLSSDAPRRIEESAIDEVVVTNTIPHEVQKLQCPKIKTVDISMILSEAIRRIHNGESMSYLFRNIGLDD | |
RUO | |
PRPSAP2 | |
Wheat Germ (in vitro) | |
GST |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.