Learn More
Abnova™ Human PRPSAP2 Full-length ORF (NP_002758.1, 1 a.a. - 369 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_002758.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5636 |
Molecular Weight (g/mol) | 67.3kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-958-372
|
Abnova™
H00005636P01S |
10 ug |
Each for $499.00
|
|
|||||
89-958-373
|
Abnova™
H00005636P01L |
25 ug |
Each for $769.00
|
|
|||||
Description
The enzyme phosphoribosylpyrophosphate synthetase (PRS) catalyzes the formation of phosphoribosylpyrophosphate which is a substrate for synthesis of purine and pyrimidine nucleotides, histidine, tryptophan and NAD. PRS exists as a complex with two catalytic subunits and two associated subunits. This gene encodes a non-catalytic associated subunit of PRS. [provided by RefSeq]
Sequence: MFCVTPPELETKMNITKGGLVLFSANSNSSCMELSKKIAERLGVEMGKVQVYQEPNRETRVQIQESVRGKDVFIIQTVSKDVNTTIMELLIMVYACKTSCAKSIIGVIPYFPYSKQCKMRKRGSIVSKLLASMMCKAGLTHLITMDLHQKEIQGFFNIPVDNLRASPFLLQYIQEEIPDYRNAVIVAKSPASAKRAQSFAERLRLGIAVIHGEAQDAESDLVDGRHSPPMVRSVAAIHPSLEIPMLIPKEKPPITVVGDVGGRIAIIVDDIIDDVDSFLAAAETLKERGAYKIFVMATHGLLSSDAPRRIEESAIDEVVVTNTIPHEVQKLQCPKIKTVDISMILSEAIRRIHNGESMSYLFRNIGLDDSpecifications
NP_002758.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
67.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MFCVTPPELETKMNITKGGLVLFSANSNSSCMELSKKIAERLGVEMGKVQVYQEPNRETRVQIQESVRGKDVFIIQTVSKDVNTTIMELLIMVYACKTSCAKSIIGVIPYFPYSKQCKMRKRGSIVSKLLASMMCKAGLTHLITMDLHQKEIQGFFNIPVDNLRASPFLLQYIQEEIPDYRNAVIVAKSPASAKRAQSFAERLRLGIAVIHGEAQDAESDLVDGRHSPPMVRSVAAIHPSLEIPMLIPKEKPPITVVGDVGGRIAIIVDDIIDDVDSFLAAAETLKERGAYKIFVMATHGLLSSDAPRRIEESAIDEVVVTNTIPHEVQKLQCPKIKTVDISMILSEAIRRIHNGESMSYLFRNIGLDD | |
RUO | |
PRPSAP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
5636 | |
PRPSAP2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC117304/MGC126719/MGC126721/PAP41 | |
PRPSAP2 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.